MilliporeSigma
  • Free radical reaction between 2-benzoyl-1,4-benzoquinones and 1,3-dicarbonyl compounds.
Free radical reaction between 2-benzoyl-1,4-benzoquinones and 1,3-dicarbonyl compounds.
Organic & biomolecular chemistry (2009-09-19)
Kuang-Po Chen, Hen-Qun Lee, Yu-Chih Cheng, Che-Ping Chuang
ABSTRACT
A manganese(III)-mediated reaction between 2-benzoyl-1,4-benzoquinones and 1,3-dicarbonyl compounds that produces benzo[c]furan-4,7-diones and anthracene-1,4-diones with high chemoselectivity is described. With ethyl butyrylacetate, by changing the solvent, benzo[c]furan-4,7-diones and anthracene-1,4-diones can be generated in high chemoselectivities. With ethyl benzoylacetate, N ,N-dimethyl acetoacetamide and 1,3-diones, benzo[c]furan-4,7-diones were produced effectively with high selectivity. With 2-alkyl-5-benzoyl-1,4-benzoquinones, the regioselectivity of this reaction was also studied and the corresponding benzo[c]furan-4,7-dione and anthracene-1,4-dione derivatives were obtained in high regioselectivity.
Sigma-Aldrich
Ethyl benzoylacetate, technical grade, 90%