MilliporeSigma
  • Phylomark, a tool to identify conserved phylogenetic markers from whole-genome alignments.
Phylomark, a tool to identify conserved phylogenetic markers from whole-genome alignments.
Applied and environmental microbiology (2012-05-15)
Jason W Sahl, Malcolm N Matalka, David A Rasko
ABSTRACT
The sequencing and analysis of multiple housekeeping genes has been routinely used to phylogenetically compare closely related bacterial isolates. Recent studies using whole-genome alignment (WGA) and phylogenetics from >100 Escherichia coli genomes has demonstrated that tree topologies from WGA and multilocus sequence typing (MLST) markers differ significantly. A nonrepresentative phylogeny can lead to incorrect conclusions regarding important evolutionary relationships. In this study, the Phylomark algorithm was developed to identify a minimal number of useful phylogenetic markers that recapitulate the WGA phylogeny. To test the algorithm, we used a set of diverse draft and complete E. coli genomes. The algorithm identified more than 100,000 potential markers of different fragment lengths (500 to 900 nucleotides). Three molecular markers were ultimately chosen to determine the phylogeny based on a low Robinson-Foulds (RF) distance compared to the WGA phylogeny. A phylogenetic analysis demonstrated that a more representative phylogeny was inferred for a concatenation of these markers compared to all other MLST schemes for E. coli. As a functional test of the algorithm, the three markers (genomic guided E. coli markers, or GIG-EM) were amplified and sequenced from a set of environmental E. coli strains (ECOR collection) and informatically extracted from a set of 78 diarrheagenic E. coli strains (DECA collection). In the instances of the 40-genome test set and the DECA collection, the GIG-EM system outperformed other E. coli MLST systems in terms of recapitulating the WGA phylogeny. This algorithm can be employed to determine the minimal marker set for any organism that has sufficient genome sequencing.
Sigma-Aldrich
Diffinity RapidTip®, for PCR Purification