MilliporeSigma
  • Synthesis of poly-(3-hydroxybutyrate-co-12 mol % 3-hydroxyvalerate) by Bacillus cereus FB11: its characterization and application as a drug carrier.
Synthesis of poly-(3-hydroxybutyrate-co-12 mol % 3-hydroxyvalerate) by Bacillus cereus FB11: its characterization and application as a drug carrier.
Journal of materials science. Materials in medicine (2013-05-16)
Farha Masood, P Chen, Tariq Yasin, Fariha Hasan, Bashir Ahmad, Abdul Hameed
ABSTRACT
The synthesis of microbial polyhydroxyalkanoate is investigated in this work for it potential application as drug carrier for cancer therapy. The bacterial isolate Bacillus cereus FB11 has synthesized poly-(3-hydroxybutyrate-co-3-hydroxyvalerate) copolymer under nutrient stress conditions using glucose as a sole carbon source. The FTIR spectrum of the purified copolymer showed the characteristic absorption bands at 1,719, 1,260 and 2,931 cm(-1) attributing to C=O, C-O stretching and C-H vibrations, respectively. The result of (1)H-NMR confirmed that it was composed of 88 mol % of 3-hydroxybutyrate and 12 mol % of 3-hydroxyvalerate monomeric subunits. The nanoparticles were fabricated from copolymer and used as a carrier for anticancer drug ellipticine. The in vitro drug release studies showed that % inhibition of A549 cancer cell line receiving ellipticine loaded poly-(3-hydroxybutyrate-co-3-hydroxyvalerate) nanoparticles was two-fold higher in comparison to ellipticine alone. This drug delivery system offers exciting possibilities for cancer therapy by increasing the bioavailability of anti-neoplastic drug to the tumor site.
Sigma-Aldrich
Diffinity RapidTip®, for PCR Purification