MilliporeSigma
  • A practical approach to new (5Z) 2-alkylthio-5-arylmethylene-1-methyl-1,5-dihydro-4H-imidazol-4-one derivatives.
A practical approach to new (5Z) 2-alkylthio-5-arylmethylene-1-methyl-1,5-dihydro-4H-imidazol-4-one derivatives.
Molecules (Basel, Switzerland) (2011-09-01)
Khadidja Bourahla, Ludovic Paquin, Olivier Lozach, Laurent Meijer, François Carreaux, Jean Pierre Bazureau
ABSTRACT
A practical protocol for the preparation of (5Z)-2-alkylthio-5-arylmethylene-1-methyl-1,5-dihydro-4H-imidazol-4-one derivatives is reported. The new compounds were obtained in good yield and stereoselectivity in two steps, namely a solvent-free Knoevenagel condensation under microwave irradiation, followed by an S-alkylation reaction with various halogenoalkanes.