MilliporeSigma
  • Degradation of 3,4-dichloro- and 3,4-difluoroaniline by Pseudomonas fluorescens 26-K.
Degradation of 3,4-dichloro- and 3,4-difluoroaniline by Pseudomonas fluorescens 26-K.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2003-03-06)
Vassili M Travkin, Inna P Solyanikova, Ivonne M C M Rietjens, Jacques Vervoort, Willem J H van Berkel, Ludmila A Golovleva
ABSTRACT
3,4-Dichloro- and 3,4-difluoroanilines were degraded by Pseudomonas fluorescens 26-K under aerobic conditions. In the presence of glucose strain degraded 170 mg/L of 3,4-dichloroaniline (3,4-DCA) during 2-3 days. Increasing of toxicant concentration up to 250 mg/L led to degradation of 3,4-DCA during 4 days and its intermediates during 5-7 days. Without cosubstrate and nitrogen source degradation of 3,4-DCA took place too, but more slowly--about 40% of toxicant at initial concentration 75 mg/L was degraded during 15 days. 3,4-Difluoroaniline (3,4-DFA) (initial concentration 170 mg/L) was degraded by Pseudomonas fluorescens 26-K during 5-7 days. The strain was able to completely degrade up to 90 mg/L of 3,4-DFA, without addition of cosubstrate and nitrogen during 15 days. Degradation of fluorinated aniline was accompanied by intensive defluorination. Activity of catechol 2,3-dioxygenase (C2,3DO) (0.230 micromol/min/mg of protein) was found in the culture liquid of the strain, grown with 3,4-DCA and glucose. This fact, as well as, the presence of 3-chloro-4-hydroxyaniline as a metabolite suggested that 3,4-DCA degradation pathway includes dehalogenation and hydroxylation of aromatic ring followed by its subsequent cleaving by C2,3DO. On the contrary, activity of catechol 1,2-dioxygenase (C1,2DO) (0.08 micromol/min/mg of protein) was found in the cell-free extract of biomass grown on 3,4-DFA. 3-Fluoro-4-hydroxyaniline as intermediate was found in this cell-free extract.
Sigma-Aldrich
3,4-Difluoroaniline, 99%