MilliporeSigma
  • Synthesis and theoretical studies on the vibration of a novel (3,4-disfluoro)phenylquione.
Synthesis and theoretical studies on the vibration of a novel (3,4-disfluoro)phenylquione.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-03-10)
Yuan-zhi Song, An-feng Zhu, Jin-shun Lv, Gui-xia Gong, Ji-min Xie, Jian-feng Zhou, Yong Ye, Xu-dong Zhong
ABSTRACT
A novel (3,4-disfluoro)phenylquione (2F-PQ) was synthesized through the reaction of 3,4-Difluoroaniline and 1,4-benzoquinone. Its structure was verified by (1)H NMR, FTIR and Raman spectra. The ground-state geometries were optimized by using density functional theory (DFT) at B3LYP/6-311G+(d,p), B3PW91/6-311G+(d,p) and MPB3PW91/6-311G+(d,p) level without symmetry constrains, respectively. The predicted FTIR and Raman spectra scaled by factor are well consistent with experimental spectra.
Sigma-Aldrich
3,4-Difluoroaniline, 99%