MilliporeSigma
  • Biosorption of heavy metals on Aspergillus fumigatus immobilized Diaion HP-2MG resin for their atomic absorption spectrometric determinations.
Biosorption of heavy metals on Aspergillus fumigatus immobilized Diaion HP-2MG resin for their atomic absorption spectrometric determinations.
Talanta (2008-10-31)
Mustafa Soylak, Mustafa Tuzen, Durali Mendil, Ibrahim Turkekul
ABSTRACT
A solid phase extraction procedure based on biosorption of copper(II), lead(II), zinc(II), iron(III), nickel(II) and cobalt(II) ions on Aspergillus fumigatus immobilized Diaion HP-2MG has been investigated. The analytical conditions including amounts of A. fumigatus, eluent type, flow rates of sample and eluent solutions were examined. Good recoveries were obtained to the spiked natural waters. The influences of the concomitant ions on the retentions of the analytes were also examined. The detection limits (3sigma, N=11) were 0.30mugl(-1) for copper, 0.32mugl(-1) for iron, 0.41mugl(-1) for zinc, 0.52mugl(-1) for lead, 0.59mugl(-1) for nickel and 0.72mugl(-1) for cobalt. The relative standard deviations of the procedure were below 7%. The validation of the presented procedure is performed by the analysis of three standard reference materials (NRCC-SLRS 4 Riverine Water, SRM 1515 Apple leaves and GBW 07605 Tea). The procedure was successfully applied for the determination of analyte ions in natural waters microwave digested samples including street dust, tomato paste, black tea, etc.
Product Description
Supelco
Diaion® HP-2MG, pkg of 100 g
Pricing and availability is not currently available.