MilliporeSigma
  • Methylenetetrahydrofolate reductase (MTHFR) C677T polymorphism and age at onset of schizophrenia: no consistent evidence for an association in the Nordic population.
Methylenetetrahydrofolate reductase (MTHFR) C677T polymorphism and age at onset of schizophrenia: no consistent evidence for an association in the Nordic population.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics (2012-10-19)
Peter Saetre, Jakob Grove, Anders D Børglum, Ole Mors, Thomas Werge, Ole A Andreassen, Maria Vares, Ingrid Agartz, Lars Terenius, Erik G Jönsson
ABSTRACT
Methylenetetrahydrofolate reductase (MTHFR) is an enzyme involved in metabolic pathways of importance for nucleotide synthesis and methylation of DNA, membranes, proteins and lipids. The MTHFR gene includes a common polymorphism (rs1801133 or C677T), which is associated with enzyme activity. The T-allele of the C677T polymorphism has been associated with earlier age at onset of schizophrenia in a Scandinavian population, although no association was found in replication attempts in other populations. Extending the study to five Nordic samples consisting of 2,198 patients with schizophrenia, including the original Scandinavian samples, there was no significant association between MTHFR C677T polymorphism and age at onset in schizophrenia. The present results do not suggest that the investigated MTHFR polymorphism has any significant influence on age at onset of schizophrenia in the Nordic population.
Sigma-Aldrich
Extract-N-Amp Blood PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications