MilliporeSigma
  • A study on stack configuration of continuous electrodeionization for removal of heavy metal ions from the primary coolant of a nuclear power plant.
A study on stack configuration of continuous electrodeionization for removal of heavy metal ions from the primary coolant of a nuclear power plant.
Water research (2004-03-18)
Kyeong-Ho Yeon, Jung-Hoon Song, Seung-Hyeon Moon
ABSTRACT
This study investigated the production of high-purity water in the primary coolant of a nuclear power plant via the continuous electrodeionization (CEDI) process, using ion exchange resins as ion-conducting media between ion exchange membranes. The effectiveness of this method was examined with respect to the removal of heavy metals. The study was carried out on a laboratory scale with an effective area of 20 cm(2). The CEDI system was operated with a layered bed of cation exchange resins, anion exchange resins, and mixed-bed ion exchange resins. The stack configuration was designed to prevent a reaction between metal ions and hydroxide ions. The CEDI operation with the layered bed removed more than 99% of the ions at 30% of the current efficiency. The results showed that, with an inlet conductivity of 40 microScm(-1), a linear velocity of 4.17 cms(-1), and an applied current density of 17 mAcm(-2), the CEDI process yielded an outlet conductivity of 0.5 microScm(-1), thereby preventing the precipitation of metal ions. This study therefore successfully demonstrated the feasibility of the CEDI operation for the removal of heavy metals at a very low concentration.