MilliporeSigma
  • Tobacco rattle virus-based virus-induced gene silencing in Nicotiana benthamiana.
Tobacco rattle virus-based virus-induced gene silencing in Nicotiana benthamiana.
Nature protocols (2014-06-06)
Muthappa Senthil-Kumar, Kirankumar S Mysore
ABSTRACT
Tobacco rattle virus (TRV)-based virus-induced gene silencing (VIGS) is widely used in various plant species to downregulate the expression of a target plant gene. TRV is a bipartite, positive-strand RNA virus with the TRV1 and TRV2 genomes. To induce post-transcriptional gene silencing (PTGS), the TRV2 genome is genetically modified to carry a fragment of the target gene and delivered into the plant (along with the TRV1 genome) by agroinoculation. TRV1- and TRV2-carrying Agrobacterium strains are then co-inoculated into 3-week-old plant leaves by one of three methods: a needleless syringe, the agrodrench method or by pricking with a toothpick. Target gene silencing occurs in the newly developed noninoculated leaves within 2-3 weeks of TRV inoculation. The TRV-VIGS protocol described here takes only 4 weeks to implement, and it is faster and easier to perform than other gene silencing techniques that are currently available. Although we use Nicotiana benthamiana as an example, the protocol is adaptable to other plant species.
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, For Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems