MilliporeSigma
  • 1,2,5-Thiadiazole derivatives are potent and selective ligands at human 5-HT1A receptors.
1,2,5-Thiadiazole derivatives are potent and selective ligands at human 5-HT1A receptors.
Bioorganic & medicinal chemistry letters (2001-05-01)
A L Sabb, R L Vogel, M G Kelly, Y Palmer, D L Smith, T H Andree, L E Schechter
ABSTRACT
Amino acid derivatives of 1,2,5-thiadiazol-3-yl-piperazine related to (+)-WAY-100135 and WAY-100635 are potent 5-HT1A receptor agonists and antagonists, which have selective affinity for 5-HT1A receptors versus alpha1 and dopamine (D2, D3, and D4) receptors.
Sigma-Aldrich
3,4-Dichloro-1,2,5-thiadiazole, 97%