MilliporeSigma
  • Antibody-mediated targeting of the Orai1 calcium channel inhibits T cell function.
Antibody-mediated targeting of the Orai1 calcium channel inhibits T cell function.
PloS one (2014-01-01)
Jennifer H Cox, Scott Hussell, Henrik Søndergaard, Kirstine Roepstorff, John-Vu Bui, Jen Running Deer, Jun Zhang, Zhan-Guo Li, Kasper Lamberth, Peter Helding Kvist, Søren Padkjær, Claus Haase, Stefan Zahn, Valerie H Odegard
ABSTRACT
Despite the attractiveness of ion channels as therapeutic targets, there are no examples of monoclonal antibodies directed against ion channels in clinical development. Antibody-mediated inhibition of ion channels could offer a directed, specific therapeutic approach. To investigate the potential of inhibiting ion channel function with an antibody, we focused on Orai1, the pore subunit of the calcium channel responsible for store-operated calcium entry (SOCE) in T cells. Effector T cells are key drivers of autoimmune disease pathogenesis and calcium signaling is essential for T cell activation, proliferation, and cytokine production. We show here the generation of a specific anti-human Orai1 monoclonal antibody (mAb) against an extracellular loop of the plasma membrane-spanning protein. The anti-Orai1 mAb binds native Orai1 on lymphocytes and leads to cellular internalization of the channel. As a result, T cell proliferation, and cytokine production is inhibited in vitro. In vivo, anti-Orai1 mAb is efficacious in a human T cell-mediated graft-versus host disease (GvHD) mouse model. This study demonstrates the feasibility of antibody-mediated inhibition of Orai1 function and, more broadly, reveals the possibility of targeting ion channels with biologics for the treatment of autoimmunity and other diseases.
Sigma-Aldrich
Anti-ORAI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution