MilliporeSigma
  • A novel technique that enables efficient conduct of simultaneous isomerization and fermentation (SIF) of xylose.
A novel technique that enables efficient conduct of simultaneous isomerization and fermentation (SIF) of xylose.
Applied biochemistry and biotechnology (2008-04-19)
Kripa Rao, Silpa Chelikani, Patricia Relue, Sasidhar Varanasi
ABSTRACT
Of the sugars recovered from lignocellulose, D-glucose can be readily converted into ethanol by baker's or brewer's yeast (Saccharomyces cerevisiae). However, xylose that is obtained by the hydrolysis of the hemicellulosic portion is not fermentable by the same species of yeasts. Xylose fermentation by native yeasts can be achieved via isomerization of xylose to its ketose isomer, xylulose. Isomerization with exogenous xylose isomerase (XI) occurs optimally at a pH of 7-8, whereas subsequent fermentation of xylulose to ethanol occurs at a pH of 4-5. We present a novel scheme for efficient isomerization of xylose to xylulose at conditions suitable for the fermentation by using an immobilized enzyme system capable of sustaining two different pH microenvironments in a single vessel. The proof-of-concept of the two-enzyme pellet is presented, showing conversion of xylose to xylulose even when the immobilized enzyme pellets are suspended in a bulk solution whose pH is sub-optimal for XI activity. The co-immobilized enzyme pellets may prove extremely valuable in effectively conducting "simultaneous isomerization and fermentation" (SIF) of xylose. To help further shift the equilibrium in favor of xylulose formation, sodium tetraborate (borax) was added to the isomerization solution. Binding of tetrahydroxyborate ions to xylulose effectively reduces the concentration of xylulose and leads to increased xylose isomerization. The formation of tetrahydroxyborate ions and the enhancement in xylulose production resulting from the complexation was studied at two different bulk pH values. The addition of 0.05 M borax to the isomerization solution containing our co-immobilized enzyme pellets resulted in xylose to xylulose conversion as high as 86% under pH conditions that are suboptimal for XI activity. These initial findings, which can be optimized for industrial conditions, have significant potential for increasing the yield of ethanol from xylose in an SIF approach.
Sigma-Aldrich
Glucose Isomerase from Streptomyces murinus, ≥350 U/g