MilliporeSigma
  • In vitro inhibitory effects of Limonium contortirameum and L. virgatum extracts from sardinia on alpha-amylase, alpha-glucosidase and pancreatic lipase.
In vitro inhibitory effects of Limonium contortirameum and L. virgatum extracts from sardinia on alpha-amylase, alpha-glucosidase and pancreatic lipase.
Natural product communications (2014-04-03)
Marzia Foddai, Violet Kasabri, Giacomo L Petretto, Emanuela Azara, Angela Sias, Fatma U Afifi, Giovanna Delogu, Mario Chessa, Giorgio Pintore
ABSTRACT
Pancreatic triacylglycerol lipase (PL), alpha-amylase and alpha-glucosidase are interesting pharmacological targets for the management of dyslipidemia, atherosclerosis, and obesity-diabetes. Limonium spp (Plumbaginaceae) are endemic to Sardinia, Italy. Comparable with acarbose, aqueous extracts (AE) of L. contortirameum and L. virgatum, and their phytoconstituent gallic acid concentration gradients (mg/mL) were identified as in vitro potent (p<0.001, n=3) and efficacious dual inhibitors of alpha-amylase and alpha-glucosidase with respective IC50 (mg/mL) values of 0.6 +/- 0.1, 1.2 +/- 0.1 and 0.15 +/- 0.02. Equivalent to orlistat (PL IC50 of 0.114 +/- 0.004 microg/mL), L. contortirameum, L. virgatum AE and their phytoprinciple gallic acid inhibited PL substantially (p<0.001, n=3) in a dose-dependent manner in vitro with PL- IC50 (microg/mL) of 920.4 +/- 105.2, 593.1 +/- 56.8 and 8.4 +/- 0.9, respectively. LC-MS analysis of extracts revealed the presence of several phenolic compounds in their aglycon and glycoside forms. These are catechins, flavones, epigallocatechins and flavonols. Flavonoid- and polyphenol-rich L contortirameum and L. virgatum, modulating gastrointestinal carbohydrate and lipid digestion and absorption, may be advocated as candidates for obesity-diabetes prevention and phytotherapy.
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Glucosidase from Bacillus stearothermophilus, lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid