MilliporeSigma
Changing role of hyaluronidase in plastic surgery.
Plastic and reconstructive surgery (2014-01-29)
Steven H Bailey, Steven Fagien, Rod J Rohrich
ABSTRACT
Soft-tissue augmentation has become an increasingly popular option for facial rejuvenation. Hyaluronic acid fillers are part of the most rapidly expanding segment of this market, largely because of their safe drug profile and temporary nature. Despite their good safety profile, they can and do have complications ranging from superficial placement, uneven placement, granulomatous reactions, and skin necrosis. This article reviews the on- and off-label uses of hyaluronidase and presents several clinical algorithms detailing the effective and safe use of hyaluronidase to manage complications secondary to hyaluronic acid fillers.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid