MilliporeSigma
  • A simple method for the determination of reduction potentials in heme proteins.
A simple method for the determination of reduction potentials in heme proteins.
FEBS letters (2014-01-21)
Igor Efimov, Gary Parkin, Elizabeth S Millett, Jennifer Glenday, Cheuk K Chan, Holly Weedon, Harpreet Randhawa, Jaswir Basran, Emma L Raven
ABSTRACT
We describe a simple method for the determination of heme protein reduction potentials. We use the method to determine the reduction potentials for the PAS-A domains of the regulatory heme proteins human NPAS2 (Em=-115 mV ± 2 mV, pH 7.0) and human CLOCK (Em=-111 mV ± 2 mV, pH 7.0). We suggest that the method can be easily and routinely applied to the determination of reduction potentials across the family of heme proteins.