MilliporeSigma
  • Three-dimensional domain swapping and supramolecular protein assembly: insights from the X-ray structure of a dimeric swapped variant of human pancreatic RNase.
Three-dimensional domain swapping and supramolecular protein assembly: insights from the X-ray structure of a dimeric swapped variant of human pancreatic RNase.
Acta crystallographica. Section D, Biological crystallography (2013-10-09)
Andrea Pica, Antonello Merlino, Alexander K Buell, Tuomas P J Knowles, Elio Pizzo, Giuseppe D'Alessio, Filomena Sica, Lelio Mazzarella
ABSTRACT
The deletion of five residues in the loop connecting the N-terminal helix to the core of monomeric human pancreatic ribonuclease leads to the formation of an enzymatically active domain-swapped dimer (desHP). The crystal structure of desHP reveals the generation of an intriguing fibril-like aggregate of desHP molecules that extends along the c crystallographic axis. Dimers are formed by three-dimensional domain swapping. Tetramers are formed by the aggregation of swapped dimers with slightly different quaternary structures. The tetramers interact in such a way as to form an infinite rod-like structure that propagates throughout the crystal. The observed supramolecular assembly captured in the crystal predicts that desHP fibrils could form in solution; this has been confirmed by atomic force microscopy. These results provide new evidence that three-dimensional domain swapping can be a mechanism for the formation of elaborate large assemblies in which the protein, apart from the swapping, retains its original fold.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 4×cryst., ~70 U/mg (acc. to Kunitz)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 90%, powder, white, ~80 U/mg
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein