MilliporeSigma
Changes in prion replication environment cause prion strain mutation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-06-05)
Nuria Gonzalez-Montalban, Young Jin Lee, Natallia Makarava, Regina Savtchenko, Ilia V Baskakov
ABSTRACT
Interspecies prion transmission often leads to stable changes in physical and biological features of prion strains, a phenomenon referred to as a strain mutation. It remains unknown whether changes in the replication environment in the absence of changes in PrP primary structure can be a source of strain mutations. To approach this question, RNA content was altered in the course of amplification of hamster strains in serial protein misfolding cyclic amplification (sPMCAb). On adaptation to an RNA-depleted environment and then readaptation to an environment containing RNA, strain 263K gave rise to a novel PrP(Sc) conformation referred to as 263K(R+), which is characterized by very low conformational stability, high sensitivity to proteolytic digestion, and a replication rate of 10(6)-fold/PMCAb round, which exceeded that of 263K by almost 10(4)-fold. A series of PMCAb experiments revealed that 263K(R+) was lacking in brain-derived 263K material, but emerged de novo as a result of changes in RNA content. A similar transformation was also observed for strain Hyper, suggesting that this phenomenon was not limited to 263K. The current work demonstrates that dramatic PrP(Sc) transformations can be induced by changes in the prion replication environment and without changes in PrP primary structure.
Sigma-Aldrich
Proteinase K from Tritirachium album, Reagents designed and manufactured under current ISO 13485 certification.
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥3.0 unit/mg solid, lyophilized powder
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, for molecular biology, ≥800 units/mL
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology