MilliporeSigma
  • Glucose-6-phosphate dehydrogenase from Saccharomyces cerevisiae is a glycoprotein.
Glucose-6-phosphate dehydrogenase from Saccharomyces cerevisiae is a glycoprotein.
Biochemical and biophysical research communications (1995-11-22)
K E Reilly, J B Allred
ABSTRACT
A commercial preparation of glucose-6-phosphate dehydrogenase (G6PD) purified from Saccharomyces cerevisiae was subjected to PAGE analysis under both nondenaturing and denaturing conditions. The enzyme, identified by both activity staining and anti-yeast G6PD antibody immunoblotting, was shown to contain carbohydrate using the highly specific periodate-digoxigenin antidigoxigenin method which is diagnostic for glycoproteins.
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)