MilliporeSigma
  • Adducts of thianthrene- and phenoxathiin cation radical tetrafluoroborates to 1-alkynes. Structures and formation of 1-(5-thianthreniumyl)- and 1-(10-phenoxathiiniumyl)alkynes on alumina leading to alpha-ketoylides and alpha-ketols.
Adducts of thianthrene- and phenoxathiin cation radical tetrafluoroborates to 1-alkynes. Structures and formation of 1-(5-thianthreniumyl)- and 1-(10-phenoxathiiniumyl)alkynes on alumina leading to alpha-ketoylides and alpha-ketols.
The Journal of organic chemistry (2005-11-19)
Paramashivappa Rangappa, Henry J Shine, John N Marx, Teyeb Ould-Ely, Anna T Kelly, Kenton H Whitmire
ABSTRACT
[structure: see text] Thianthrene cation radical tetrafluoroborate (Th*+ BF4(-)) added to the terminal alkynes 1-pentyne, 1-hexyne, 1-heptyne, 1-octyne, 1-nonyne, and 1-decyne to form trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborates (1-6). Similarly, addition of phenoxathiin cation radical tetrafluoroborate (PO*+ BF4(-)) to the same alkynes gave 1,2-bis(10-phenoxathiiniumyl)alkene tetrafluoroborates (7-12). The trans configuration of two of the adducts (1 and 4) was shown with X-ray crystallography. When solutions of 1-6 in chloroform were stirred with activated alumina, cis elimination of a proton and thianthrene (Th) occurred with the formation of 1-(5-thianthreniumyl)alkyne tetrafluoroborates (1a-6a). Similar treatment of 8-12 caused elimination of a proton and phenoxathiin (PO) with formation of 1-(10-phenoxathiiniumyl)alkene tetrafluoroborates (8a-12a). Stirring of 1a-6a with alumina for short periods of time caused their conversion into 5-[(alpha-keto)alkyl]thianthrenium ylides (1b-6b) and alpha-ketols, RC(O)CH2OH (1c-6c).
Sigma-Aldrich
1-Nonyne, 99%