MilliporeSigma
  • A genome-wide association study identifies GLT6D1 as a susceptibility locus for periodontitis.
A genome-wide association study identifies GLT6D1 as a susceptibility locus for periodontitis.
Human molecular genetics (2009-11-10)
Arne S Schaefer, Gesa M Richter, Michael Nothnagel, Thomas Manke, Henrik Dommisch, Gunnar Jacobs, Alexander Arlt, Philip Rosenstiel, Barbara Noack, Birte Groessner-Schreiber, Søren Jepsen, Bruno G Loos, Stefan Schreiber
ABSTRACT
Periodontitis is a widespread, complex inflammatory disease of the mouth, which results in a loss of gingival tissue and alveolar bone, with aggressive periodontitis (AgP) as its most severe form. To identify genetic risk factors for periodontitis, we conducted a genome-wide association study in German AgP patients. We found AgP to be strongly associated with the intronic SNP rs1537415, which is located in the glycosyltransferase gene GLT6D1. We replicated the association in a panel of Dutch generalized and localized AgP patients. In the combined analysis including 1758 subjects, rs1537415 reached a genome-wide significance level of P= 5.51 x 10(-9), OR = 1.59 (95% CI 1.36-1.86). The associated rare G allele of rs1537415 showed an enrichment of 10% in periodontitis cases (48.4% in comparison with 38.8% in controls). Fine-mapping and a haplotype analysis indicated that rs1537415 showed the strongest association signal. Sequencing identified no further associated variant. Tissue-specific expression analysis of GLT6D1 indicated high transcript levels in the leukocytes, the gingiva and testis. Analysis of potential transcription factor binding sites at this locus predicted a significant reduction of GATA-3 binding affinity, and an electrophoretic mobility assay indicated a T cell specific reduction of protein binding for the G allele. Overexpression of GATA-3 in HEK293 cells resulted in allele-specific binding of GATA-3, indicating the identity of GATA-3 as the binding protein. The identified association of GLT6D1 with AgP implicates this locus as an important susceptibility factor, and GATA-3 as a potential signaling component in the pathophysiology of periodontitis.