MilliporeSigma
  • Rab geranylgeranyl transferase alpha mutation in the gunmetal mouse reduces Rab prenylation and platelet synthesis.
Rab geranylgeranyl transferase alpha mutation in the gunmetal mouse reduces Rab prenylation and platelet synthesis.
Proceedings of the National Academy of Sciences of the United States of America (2000-03-29)
J C Detter, Q Zhang, E H Mules, E K Novak, V S Mishra, W Li, E B McMurtrie, V T Tchernev, M R Wallace, M C Seabra, R T Swank, S F Kingsmore
ABSTRACT
Few molecular events important to platelet biogenesis have been identified. Mice homozygous for the spontaneous, recessive mutation gunmetal (gm) have prolonged bleeding, thrombocytopenia, and reduced platelet alpha- and delta-granule contents. Here we show by positional cloning that gm results from a G-->A substitution mutation in a splice acceptor site within the alpha-subunit of Rab geranylgeranyl transferase (Rabggta), an enzyme that attaches geranylgeranyl groups to Rab proteins. Most Rabggta mRNAs from gm tissues skipped exon 1 and lacked a start codon. Rabggta protein and Rab geranylgeranyl transferase (GGTase) activity were reduced 4-fold in gm platelets. Geranylgeranylation and membrane association of Rab27, a Rab GGTase substrate, were significantly decreased in gm platelets. These findings indicate that geranylgeranylation of Rab GTPases is critical for hemostasis. Rab GGTase inhibition may represent a new treatment for thrombocytosis and clotting disorders.