MilliporeSigma
  • An N-heterocyclic carbene-catalyzed [8 + 3] annulation of tropone and enals via homoenolate.
An N-heterocyclic carbene-catalyzed [8 + 3] annulation of tropone and enals via homoenolate.
The Journal of organic chemistry (2006-11-04)
Vijay Nair, Manojkumar Poonoth, Sreekumar Vellalath, Eringathodi Suresh, Rajasekaran Thirumalai
ABSTRACT
A novel protocol for the annulation of tropone to enals involving nucleophilic heterocyclic carbene (NHC) catalyzed homoenolate formation has been developed. Interestingly, the reaction led to bicyclic delta-lactones instead of the expected gamma-spirolactones, presumably by the uncommon [8 + 3] annulation pathway. The strategy works well with a variety of enals.
Sigma-Aldrich
Tropone, 97%