MilliporeSigma
Nanochannel-based single molecule recycling.
Nano letters (2012-06-06)
John F Lesoine, Prahnesh A Venkataraman, Peter C Maloney, Mark E Dumont, Lukas Novotny
ABSTRACT
We present a method for measuring the fluorescence from a single molecule hundreds of times without surface immobilization. The approach is based on the use of electroosmosis to repeatedly drive a single target molecule in a fused silica nanochannel through a stationary laser focus. Single molecule fluorescence detected during the transit time through the laser focus is used to repeatedly reverse the electrical potential controlling the flow direction. Our method does not rely on continuous observation and therefore is less susceptible to fluorescence blinking than existing fluorescence-based trapping schemes. The variation in the turnaround times can be used to measure the diffusion coefficient on a single molecule level. We demonstrate the ability to recycle both proteins and DNA in nanochannels and show that the procedure can be combined with single-pair Förster energy transfer. Nanochannel-based single molecule recycling holds promise for studying conformational dynamics on the same single molecule in solution and without surface tethering.
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 532 maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)