MilliporeSigma
The cytotoxicity of psoralidin from Psoralea corylifolia.
Planta medica (1996-08-01)
Y M Yang, J W Hyun, M S Sung, H S Chung, B K Kim, W H Paik, S S Kang, J G Park
ABSTRACT
A cytotoxic coumestan derivative, psoralidin (1), was isolated from the seed of Psoralea corylifolia. The IC50 values of 1 against SNU-1 and SNU-16 carcinoma cell lines were 53 and 203 micrograms/ml, respectively, indicating cytotoxic activity against stomach carcinoma cell lines.
Sigma-Aldrich
Psoralidin, ≥98% (HPLC)