MilliporeSigma
Benzomalvins, new substance P inhibitors from a Penicillium sp.
The Journal of antibiotics (1994-05-01)
H H Sun, C J Barrow, D M Sedlock, A M Gillum, R Cooper
ABSTRACT
In the course of screening microbial broths for neurokinin receptor antagonists, a series of new benzodiazepines, benzomalvins A (1), B (2) and C (3), has been isolated from the culture broth of a fungus identified as a Penicillium sp. Benzomalvin A (1) showed inhibitory activity against substance P with Ki values of 12, 42 and 43 microM at the guinea pig, rat and human neurokinin NK1 receptors, respectively. Benzomalvins B (2) and C (3) were only weakly active. The structures of these compounds were determined by spectroscopic methods including MS measurements and NMR analysis.
Sigma-Aldrich
Benzomalvin-C, ≥95% (LC/MS-ELSD)