- Trichomonas infection in pregnancy--does it affect perinatal outcome?
Trichomonas infection in pregnancy--does it affect perinatal outcome?
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde (1983-04-09)
S M Ross, A van Middelkoop
PMID6845046
ABSTRACT
Antenatal patients free of Trichomonas vaginalis vaginal infection were compared with infected patients, half of whom were treated and half left untreated. The treated group was given benzoylmetronidazole 50 ml (2 g metronidazole equivalent) as a single dose. The mode of treatment was found to be very palatable and highly effective. The birth weights and gestational age at delivery were similar in all three groups.
Product Description
Metronidazole benzoate, European Pharmacopoeia (EP) Reference Standard