MilliporeSigma
  • The memory-enhancing effects of the piracetam-like nootropics are dependent on experimental parameters.
The memory-enhancing effects of the piracetam-like nootropics are dependent on experimental parameters.
Behavioural brain research (1989-05-01)
C Mondadori, T Ducret, J Borkowski
ABSTRACT
The effects of the nootropic agent piracetam and its congeners oxiracetam, pramiracetam and aniracetam on the retention performance of mice in a passive-avoidance situation are dependent on the intensity of the foot-shock applied. This phenomenon is observed upon both pre-trial and post-trial drug administration.
Sigma-Aldrich
Pramiracetam, ≥98% (HPLC)