MilliporeSigma
  • Population pharmacokinetic-pharmacodynamic modelling of mycophenolic acid in paediatric renal transplant recipients in the early post-transplant period.
Population pharmacokinetic-pharmacodynamic modelling of mycophenolic acid in paediatric renal transplant recipients in the early post-transplant period.
British journal of clinical pharmacology (2014-05-20)
Min Dong, Tsuyoshi Fukuda, Shareen Cox, Marij T de Vries, David K Hooper, Jens Goebel, Alexander A Vinks
ABSTRACT
The purpose of this study was to develop a population pharmacokinetic and pharmacodynamic (PK-PD) model for mycophenolic acid (MPA) in paediatric renal transplant recipients in the early post-transplant period. A total of 214 MPA plasma concentrations-time data points from 24 patients were available for PK model development. In 17 out of a total of 24 patients, inosine monophosphate dehydrogenase (IMPDH) enzyme activity measurements (n = 97) in peripheral blood mononuclear cells were available for PK-PD modelling. The PK-PD model was developed using non-linear mixed effects modelling sequentially by 1) developing a population PK model and 2) incorporating IMPDH activity into a PK-PD model using post hoc Bayesian PK parameter estimates. Covariate analysis included patient demographics, co-medication and clinical laboratory data. Non-parametric bootstrapping and prediction-corrected visual predictive checks were performed to evaluate the final models. A two compartment model with a transit compartment absorption best described MPA PK. A non-linear relationship between dose and MPA exposure was observed and was described by a power function in the model. The final population PK parameter estimates (and their 95% confidence intervals) were CL/F, 22 (14.8, 25.2) l h(-1) 70 kg(-1) ; Vc /F, 45.4 (29.6, 55.6) l; Vp /F, 411 (152.6, 1472.6)l; Q/F, 22.4 (16.0, 32.5) l h(-1) ; Ka , 2.5 (1.45, 4.93) h(-1) . Covariate analysis in the PK study identified body weight to be significantly correlated with CL/F. A simplified inhibitory Emax model adequately described the relationship between MPA concentration and IMPDH activity. The final population PK-PD parameter estimates (and their 95% confidence intervals) were: E0 , 3.45 (2.61, 4.56) nmol h(-1)  mg(-1) protein and EC50 , 1.73 (1.16, 3.01) mg l(-1) . Emax was fixed to 0. There were two African-American patients in our study cohorts and both had low IMPDH baseline activities (E0 ) compared with Caucasian patients (mean value 2.13 mg l(-1) vs. 3.86 mg l(-1) ). An integrated population PK-PD model of MPA has been developed in paediatric renal transplant recipients. The current model provides information that will facilitate future studies and may be implemented in a Bayesian algorithm to allow a PK-PD guided therapeutic drug monitoring strategy.
Product Description
Supelco
Propofol-D17 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®