MilliporeSigma
  • The many faces of rare diseases (RD): meeting report on Rare Disease in South-Eastern Europe, 15-16 November 2013, Skopje, Republic of Macedonia.
The many faces of rare diseases (RD): meeting report on Rare Disease in South-Eastern Europe, 15-16 November 2013, Skopje, Republic of Macedonia.
Pediatric endocrinology reviews : PER (2014-04-11)
Z Gucev, V Tasic, M Polenakovic
ABSTRACT
The Second meeting on Rare Diseases in South Eastern Europe (SEE) was held in Skope, Macedonia on November 15-16, 2013. Objective and main data: Rare diseases (RD) are a major problem in developed and especially in countries without affluence. 6-8% of every population suffers from RD. The cumulative effect of RDs on the health system of a country is increasing. Diagnosis often remains a challenge and requires international collaboration. Treatment in diseases for which medication exist is often inaccessible to patients because of the high costs. All countries of SEE need screening programs that address more diseases. Patient organizations play a major role in increasing awareness and providing the needed pressure on society to treat treatable RDs. On the other hand, RDs are frequently a source of valuable new molecular insights not only on mechanisms of their etiology and pathology, but sometimes provide an insight on mechanisms of frequent diseases in man. Further efforts are needed in improving all the RD aspects mentioned.
Product Description
Somatropin, European Pharmacopoeia (EP) Reference Standard