MilliporeSigma
  • Combined administration of gonadotropin-releasing hormone agonist with human chorionic gonadotropin for final oocyte maturation in GnRH antagonist cycles for in vitro fertilization.
Combined administration of gonadotropin-releasing hormone agonist with human chorionic gonadotropin for final oocyte maturation in GnRH antagonist cycles for in vitro fertilization.
The Journal of reproductive medicine (2014-03-07)
Chung-Hoon Kim, Jun-Woo Ahn, Rae-Mi You, Sung-Hoon Kim, Hee-Dong Chae, Byung-Moon Kang
ABSTRACT
To evaluate the effects of combined administration of gonadotropin-releasing hormone (GnRH) agonist with human chorionic gonadotropin (hCG)for final oocyte maturation in GnRH antagonist cycles for in vitro fertilization (IVF). A total of 120 infertile women undergoing GnRH antagonist multiple-dose protocol for controlled ovarian stimulation were recruited and randomized into 2 groups: a study group (n=60) and a control group (n=60). For the study group, both GnRH agonist and recombinant hCG (rhCG) were injected concomitantly for final oocyte maturation when 1 or more follicles reached a mean diameter of 18 mm. For the control group, rhCG alone was administered for final oocyte maturation. There were no significant differences in patient characteristics. The 2 groups were also similar with respect to the number of oocytes retrieved, fertilized oocytes and good-quality embryos. Embryo implantation rate (24.7% vs. 14.9%), clinical pregnancy rate per cycle (53.3% vs. 33.3%), and live birth rate (50.0% vs. 30.0%) were significantly higher in the study group than in the control group (p = 0.006, p = 0.027, and p = 0.025, respectively). Combined administration of GnRH agonist with rhCG may be beneficial in improving endometrial receptivity and pregnancy rate in GnRH antagonist cycles for IVF.
Product Description
Gonadorelin, European Pharmacopoeia (EP) Reference Standard