MilliporeSigma
  • Predictors of asthma control and lung function responsiveness to step 3 therapy in children with uncontrolled asthma.
Predictors of asthma control and lung function responsiveness to step 3 therapy in children with uncontrolled asthma.
The Journal of allergy and clinical immunology (2013-10-03)
Nathan Rabinovitch, David T Mauger, Nichole Reisdorph, Ronina Covar, Jonathan Malka, Robert F Lemanske, Wayne J Morgan, Theresa W Guilbert, Robert S Zeiger, Leonard B Bacharier, Stanley J Szefler
ABSTRACT
Predictors of improvement in asthma control and lung function to step 3 therapy in children with persistent asthma have not been identified despite reported heterogeneity in responsiveness. We sought to evaluate potential predictors of asthma control and lung function responsiveness to step 3 therapy. A post hoc analysis from the Best Add-On Giving Effective Response (BADGER) study tested the association between baseline biological, asthma control, pulmonary function, and demographic markers and responsiveness to step-up to a higher dose of inhaled corticosteroid (ICS step-up therapy) or addition of leukotriene receptor antagonist (LTRA step-up therapy) or long-acting β₂-agonist (LABA step-up therapy). In multivariate analyses higher impulse oscillometry reactance area was associated (P = .048) with a differential FEV₁ response favoring LABA over ICS step-up therapy, whereas higher urinary leukotriene E₄ levels were marginally (P = .053) related to a differential FEV₁ response favoring LTRA over LABA step-up therapy. Predictors of differential responses comparing ICS with LTRA step-up therapy were not apparent, probably because of suppression of allergic markers with low-dose ICS treatment. Minimal overlap was seen across FEV₁ and asthma control day predictors, suggesting distinct mechanisms related to lung function and asthma control day responses. Levels of impulse oscillometry reactance area indicating peripheral airway obstruction and urinary leukotriene E₄ levels indicating cysteinyl leukotriene inflammation can differentiate LABA step-up responses from responses to LTRA or ICS step-up therapy. Further studies with physiologic, genetic, and biological markers related to these phenotypes will be needed to predict individual responses to LABA step-up therapy.
Product Description
Montelukast sodium, European Pharmacopoeia (EP) Reference Standard