MilliporeSigma
  • Ovulation-inducing drugs and ovarian cancer risk: results from an extended follow-up of a large United States infertility cohort.
Ovulation-inducing drugs and ovarian cancer risk: results from an extended follow-up of a large United States infertility cohort.
Fertility and sterility (2013-09-10)
Britton Trabert, Emmet J Lamb, Bert Scoccia, Kamran S Moghissi, Carolyn L Westhoff, Shelley Niwa, Louise A Brinton
ABSTRACT
To examine the relationship of ovulation-inducing drugs and ovarian cancer. Retrospective cohort study, with additional follow-up since initial report. Five large reproductive endocrinology practices. In a retrospective cohort of 9,825 women evaluated for infertility at five clinical sites in the United States between 1965 and 1988 with follow-up through 2010, we examined the relationship of ovulation-inducing drugs and ovarian cancer (n = 85). None. Hazard rate ratios (RR) and 95% confidence intervals (CI) for ovarian cancer. Among women evaluated for infertility, there was no association of ovarian cancer risk with ever use of clomiphene citrate (CC) (adjusted RR 1.34, 95% CI 0.86-2.07) or gonadotropins (RR 1.00, 95% CI 0.48-2.08) and no evidence that any of several more detailed subgroups of usage were related to an increased risk with one exception: women who used CC and remained nulligravid did demonstrate much higher risks than those who successfully conceived compared with nonusers (respectively, RR 3.63, 95% CI 1.36-9.72 vs. RR 0.88, 95% CI 0.47-1.63). Our overall results were reassuring and consistent with other studies. A reason for an association between CC use and ovarian cancer among persistently nulligravid women remains to be determined. Given the large and increasing number of women treated with ovulation-inducing drugs, the increased risk of ovarian cancer among the subset of women who remained nulligravid should be further monitored.
Sigma-Aldrich
Enclomiphene hydrochloride, ≥96% (HPLC)