MilliporeSigma
  • Antidepressant and anticonvulsant effects of exercise in a rat model of epilepsy and depression comorbidity.
Antidepressant and anticonvulsant effects of exercise in a rat model of epilepsy and depression comorbidity.
Epilepsy & behavior : E&B (2013-08-13)
S Alisha Epps, Alexa B Kahn, Philip V Holmes, Katherine A Boss-Williams, Jay M Weiss, David Weinshenker
ABSTRACT
The bidirectional comorbidity between epilepsy and depression is associated with severe challenges for treatment efficacy and safety, often resulting in poor prognosis and outcome for the patient. We showed previously that rats selectively bred for depression-like behaviors (SwLo rats) also have increased limbic seizure susceptibility compared with their depression-resistant counterparts (SwHi rats). In this study, we examined the therapeutic efficacy of voluntary exercise in our animal model of epilepsy and depression comorbidity. We found that chronic wheel running significantly increased both struggling duration in the forced swim test and latency to pilocarpine-induced limbic motor seizure in SwLo rats but not in SwHi rats. The antidepressant and anticonvulsant effects of exercise were associated with an increase in galanin mRNA specifically in the locus coeruleus of SwLo rats. These results demonstrate the beneficial effects of exercise in a rodent model of epilepsy and depression comorbidity and suggest a potential role for galanin.