MilliporeSigma
  • A 24-week, randomized, controlled trial of rivastigmine patch 13.3 mg/24 h versus 4.6 mg/24 h in severe Alzheimer's dementia.
A 24-week, randomized, controlled trial of rivastigmine patch 13.3 mg/24 h versus 4.6 mg/24 h in severe Alzheimer's dementia.
CNS neuroscience & therapeutics (2013-08-09)
Martin R Farlow, George T Grossberg, Carl H Sadowsky, Xiangyi Meng, Monique Somogyi
ABSTRACT
The 24-week, prospective, randomized, double-blind ACTION study investigated the efficacy, safety, and tolerability of 13.3 versus 4.6 mg/24 h rivastigmine patch in patients with severe Alzheimer's disease (AD). Patients had probable AD and Mini-Mental State Examination scores ≥3-≤12. Primary outcome measures were as follows: Severe Impairment Battery (SIB) and AD Cooperative Study-Activities of Daily Living scale-Severe Impairment Version (ADCS-ADL-SIV). Secondary outcomes were as follows: ADCS-Clinical Global Impression of Change (ADCS-CGIC), 12-item Neuropsychiatric Inventory (NPI-12), and safety/tolerability. Of 1014 patients screened, 716 were randomized to 13.3 mg/24 h (N = 356) or 4.6 mg/24 h (N = 360) patch. Baseline characteristics/demographics were comparable. Completion rates were as follows: 64.3% (N = 229) with 13.3 mg/24 h and 65.0% (N = 234) with 4.6 mg/24 h patch. The 13.3 mg/24 h patch was significantly superior to 4.6 mg/24 h patch on cognition (SIB) and function (ADCS-ADL-SIV) at Week 16 (P < 0.0001 and P = 0.049, respectively) and 24 (primary endpoint; P < 0.0001 and P = 0.025). Significant between-group differences (Week 24) were observed on the ADCS-CGIC (P = 0.0023), not NPI-12 (P = 0.1437). A similar proportion of the 13.3 mg/24 h and 4.6 mg/24 h patch groups reported adverse events (AEs; 74.6% and 73.3%, respectively) and serious AEs (14.9% and 13.6%). The 13.3 mg/24 h patch demonstrated superior efficacy to 4.6 mg/24 h patch on SIB and ADCS-ADL-SIV, without marked increase in AEs, suggesting higher-dose patch has a favorable benefit-to-risk profile in severe AD.
Sigma-Aldrich
Rivastigmine tartrate, ≥98% (HPLC)
Rivastigmine for system suitability, European Pharmacopoeia (EP) Reference Standard
Rivastigmine hydrogen tartrate, European Pharmacopoeia (EP) Reference Standard