MilliporeSigma
  • CTX-M-123, a novel hybrid of the CTX-M-1 and CTX-M-9 Group β-lactamases recovered from Escherichia coli isolates in China.
CTX-M-123, a novel hybrid of the CTX-M-1 and CTX-M-9 Group β-lactamases recovered from Escherichia coli isolates in China.
Antimicrobial agents and chemotherapy (2013-06-12)
Dandan He, Sally R Partridge, Jianzhong Shen, Zhenling Zeng, Lanping Liu, Lili Rao, Luchao Lv, Jian-Hua Liu
ABSTRACT
The chimeric bla(CTX-M-123) gene was identified in two ceftazidime-resistant Escherichia coli isolates from animals in different Chinese provinces. Like other CTX-M-1/9 group hybrids (CTX-M-64 and CTX-M-132), the ends (amino acids 1 to 135 and 234 to 291) of CTX-M-123 match CTX-M-15 while the central part (122 to 241) matches CTX-M-14. bla(CTX-M-123) is carried on related, but not identical, ~90-kb IncI1 plasmids in the two isolates, and one isolate simultaneously carries the group 1 blaCTX-M-55 gene on an additional IncI2 plasmid.
Product Description
Ceftazidime for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ceftazidime pentahydrate, United States Pharmacopeia (USP) Reference Standard
Ceftazidime, European Pharmacopoeia (EP) Reference Standard