MilliporeSigma
  • Analgesic effect of a single preoperative dose of the antibiotic ceftriaxone in humans.
Analgesic effect of a single preoperative dose of the antibiotic ceftriaxone in humans.
The journal of pain : official journal of the American Pain Society (2013-06-04)
Angela Macaluso, Matteo Bernabucci, Angela Trabucco, Ludovico Ciolli, Fabiana Troisi, Rossella Baldini, Roberto Gradini, Giuseppe Battaglia, Ferdinando Nicoletti, Saul Collini
ABSTRACT
Repeated injections of the antibiotic ceftriaxone cause analgesia in rodents by upregulating the glutamate transporter, GLT-1. No evidence is available in humans. We studied the effect of a single intravenous administration of ceftriaxone in patients undergoing decompressive surgery of the median or ulnar nerves. Forty-five patients were randomized to receive saline, ceftriaxone (2 g), or cefazolin (2 g), 1 hour before surgery. Cefazolin, which is structurally related to ceftriaxone, was used as a negative control. Pain thresholds were measured 10 minutes before drug injections and then 4 to 6 hours after surgery. Ceftriaxone caused analgesia in all patients, whereas cefazolin was inactive. We also performed animal studies to examine whether a single dose of ceftriaxone was sufficient to induce analgesia. A single intraperitoneal injection of ceftriaxone (200 mg/kg), but not cefazoline (200 mg/kg), caused analgesia in mouse models of inflammatory or postsurgical pain, and upregulated GLT-1 in the spinal cord. Ceftriaxone-induced analgesia was additive to that produced by blockade of mGlu5 receptors, which are activated by extrasynaptic glutamate. These data indicate that a single dose of ceftriaxone causes analgesia in humans and mice and suggest that ceftriaxone should be used for preoperative antimicrobial prophylaxis when a fast relief of pain is desired. The study reports for the first time that a single preoperative dose of ceftriaxone causes analgesia in humans. A single dose of ceftriaxone could also relieve inflammatory and postsurgical pain and upregulate GLT-1 expression in mice. Ceftriaxone should be preferred to other antibiotics for antimicrobial prophylaxis to reduce postoperative pain.
Product Description
Cefazolin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Cefazolin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.