MilliporeSigma
[Ideas about registration for breast implants].
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery (2012-10-13)
Peng Zhao, Xinli Shi, Wenbo Liu
ABSTRACT
To review the registration and technical data for breast implants. Recent literature concerning registration for breast implants was reviewed and analyzed. The aspects on registration for breast implants are complicated. The major aspects include providing technical data, establishing develop standard, progress type test, and developing the registration specification. The manufacturers need to be well aware of the risk of this kind of product and have the full ability to conduct the researching and developing. Only the strict risk controlling can ensure the safety and effectiveness of the product.
Product Description
USP
Simethicone, United States Pharmacopeia (USP) Reference Standard