MilliporeSigma
  • Fast screening for antioxidant properties of flavonoids from Pterogyne nitens using electrochemical methods.
Fast screening for antioxidant properties of flavonoids from Pterogyne nitens using electrochemical methods.
Journal of AOAC International (2012-07-24)
Leonardo Luiz Okumura, Luis Octávio Regasini, Daniara Cristina Fernandes, Dulce Helena Siqueira da Silva, Maria Valnice Boldrin Zanoni, Vanderlan da Silva Bolzani
ABSTRACT
A fast, low-cost, convenient, and especially sensitive voltammetric screening approach for the study of the antioxidant properties of isoquercitrin and pedalitin from Pterogyne nitens is suggested in this work. These flavonoids were investigated for their redox properties using cyclic voltammetry in nonaqueous media using N,N-dimethylformamide and tetrabutylammonium tetrafluorborate as the supporting electrolyte, a glassy carbon working electrode, A6(see symbol in text)AgCI reference electrode, and Pt bare wire counter electrode. The comparative analysis of the activity of rutin has also been carried out. Moreover, combining HPLC with an electrochemical detector allowed qualitative and quantitative detection of micromolecules (e.g., isoquercitrin and pedalitin) that showed antioxidant activities. These results were then correlated to the inhibition of beta-carotene bleaching determined by TLC autographic assay and to structural features of the flavonoids.
Product Description
Isoquercitroside, European Pharmacopoeia (EP) Reference Standard