MilliporeSigma
  • Leonurine attenuates lipopolysaccharide-induced inflammatory responses in human endothelial cells: involvement of reactive oxygen species and NF-κB pathways.
Leonurine attenuates lipopolysaccharide-induced inflammatory responses in human endothelial cells: involvement of reactive oxygen species and NF-κB pathways.
European journal of pharmacology (2012-02-07)
Xin Hua Liu, Li Long Pan, He Bei Yang, Qi Hai Gong, Yi Zhun Zhu
ABSTRACT
Leonurine, an active alkaloid of Traditional Chinese Medicine Herba leonuri, displayed cardioprotective effects by anti-oxidative and anti-apoptotic activities in vitro and in vivo. Herein, we explored the effects and possible mechanisms of leonurine on lipopolysaccharide (LPS)-induced inflammatory responses in human umbilical vein endothelial cells (HUVEC). We found that leonurine pretreatment concentration-dependently attenuated LPS-induced mRNA expression of intercellular adhesion molecule-1 (ICAM-1), vascular cell adhesion molecule-1 (VCAM-1), E-selectin, and monocyte chemoattractant protein-1. Meanwhile, LPS-mediated expression/release of ICAM-1, VCAM-1, and cyclooxygenase-2, and tumor necrosis factor-α was also reduced by leonurine. In addition, we confirmed that leonurine suppressed degradation of IκBα and phosphorylation of nuclear factor-κB (NF-κB) p65 as well as production of intracellular reactive oxygen species in a concentration dependent manner. Furthermore, the cytoprotective enzyme heme oxygenase-1 could be upregulated in leonurine-treated HUVEC. Our present results indicated leonurine exerted beneficial effects in inflammatory conditions partly through inhibition of reactive oxygen species and NF-κB signaling pathways.
Sigma-Aldrich
Leonurine, ≥98% (HPLC)