MilliporeSigma
  • In vivo evaluation of cisplatin-loaded superabsorbent polymer microspheres for use in chemoembolization of VX2 liver tumors.
In vivo evaluation of cisplatin-loaded superabsorbent polymer microspheres for use in chemoembolization of VX2 liver tumors.
Journal of vascular and interventional radiology : JVIR (2012-01-03)
Noboru Maeda, Keigo Osuga, Kouki Shimazu, Eiichi Morii, Koji Mikami, Shinichi Hori, Tetsuro Nakazawa, Hiroki Higashihara, Kaname Tomoda, Hironobu Nakamura, Noriyuki Tomiyama
ABSTRACT
To investigate the pharmacokinetics and efficacy of chemoembolization with a cisplatin-loaded superabsorbent polymer (SAP) suspension in a rabbit model with transplanted liver VX2 tumors. VX2 tumors were implanted into the left lobe of the liver in eight rabbits. Embolization of the proper hepatic artery was performed with cisplatin-loaded or unloaded SAP. In the cisplatin-loaded SAP group (n = 4), 5 mg of SAP (106-150 μm) loading 2.35 mg of cisplatin and 0.5 mL of ionic contrast material (ioxaglic acid 320 mgI/mL) was injected into the proper hepatic artery. In the control group (hepatic arterial infusion [HAI] + SAP; n = 4), 5 mg of SAP loading 0.5 mL of ioxaglic acid alone was injected after a bolus infusion of an equivalent amount of cisplatin. Sequential change of the plasma platinum concentration within the first 24 hours was measured. Blood sampling and histopathologic examination were performed at 1-week follow-up. Magnetic resonance (MR) images were used to calculate the growth rate of the VX2 tumor. All animals underwent successful embolization. Both total and free plasma platinum mean concentrations within the first 24 hours remained lower in the cisplatin-loaded SAP group, although without statistical significance (P > .05). The mean tumor growth rate was significantly lower in the cisplatin-loaded SAP group than the control group (20% vs 116%; P = .049). Histopathologic examination revealed coagulative necrosis to nontumorous liver parenchyma in two rabbits in the cisplatin-loaded SAP group, although no deaths occurred. These results suggested that chemoembolization with cisplatin-loaded SAP was a safe and tolerable treatment and was more effective in suppressing the tumor growth.
Product Description
USP
Ioxaglic acid, United States Pharmacopeia (USP) Reference Standard
Ioxaglic acid, European Pharmacopoeia (EP) Reference Standard