MilliporeSigma
  • Outcome of continence procedures after failed endoscopic treatment with dextranomer-based implants (DEFLUX®).
Outcome of continence procedures after failed endoscopic treatment with dextranomer-based implants (DEFLUX®).
Journal of pediatric urology (2011-02-01)
I Alova, M Margaryan, V Verkarre, M Bernuy, S Lortat Jacob, H B Lottmann
ABSTRACT
To evaluate outcome of further continence procedures after failure of endoscopic injections of dextranomer-based bulking agent. From 1997, 89 children (3-18 years) and one young adult were treated for incontinence with 145 endoscopic injections of dextranomer. On evaluation, each patient was classified as: dry, significantly improved, or treatment failure. Eighty-five patients had at least 12 months of follow up. Out of 34 (40%) treatment failures, 24 patients had a subsequent bladder neck procedure: artificial urinary sphincter (7), bladder neck plasty (9), bladder neck closure (1), fascial sling (3). Six patients had further endoscopic treatment (including 2 after bladder neck plasty). At surgery, Deflux(®) paste was easily identified with minimal surrounding tissue inflammatory reaction. Artificial urinary sphincter: all 7 dry; bladder neck plasty: 7 dry, 1 improved and 1 still incontinent; 1 bladder neck closure patient dry; fascial sling: 1 significantly improved and 2 dry. Repeated endoscopic treatment: 1 dry, 1 improved and 4 failed. Eleven of the 21 (52%) patients who are either dry or improved have voiding abilities. Endoscopic injections with dextranomer do not adversely affect the outcome of further surgical procedures. Repeated injections after a previous endoscopic treatment failure have a low success rate.
Product Description
Dextranomer, European Pharmacopoeia (EP) Reference Standard