MilliporeSigma
  • Hemisynthesis of dihydroumbellulols from umbellulone: new cooling compounds.
Hemisynthesis of dihydroumbellulols from umbellulone: new cooling compounds.
Journal of agricultural and food chemistry (2010-12-31)
Christian Starkenmann, Isabelle Cayeux, Robert Brauchli, Fabienne Mayenzet
ABSTRACT
Although menthol is a common ingredient used in food products, other molecules also evoke coolness through stimulation of the somatosensory system. To discover new molecules having cooling properties, we virtually screened the chemical structures of terpenes and sesquiterpenes to find structures that are similar to (-)-menthol. We realized that dihydroumbellulols could be good candidates. Although their occurrence was reported in Hyptis pectinata Poit, we were unable to obtain these molecules from the plant or to prove their natural occurrence. Therefore, we extracted (-)-(R)-umbellulone from Umbellularia californica Nutt. The (-)-(R)-umbellulone was reduced to prepare (1R,2R/S)-1-isopropyl-4-methylbicyclo[3.1.0]hex-3-en-2-ol, (1R,4R/S)-1-isopropyl-4-methylbicyclo[3.1.0]hexan-2-one, and (1R,2RS,4RS)-1-isopropyl-4-methylbicyclo[3.1.0]hexan-2-ols, named dihydroumbellulols. Sensory analysis suggested that (1R,2R,4S)-dihydroumbellulol has a pleasant, trigeminal cooling effect, about 2-3 times less cooling than (-)-menthol, with a weak odor slightly reminiscent of eucalyptol. In addition, a previously unreported compound was discovered, (-)-(1R)-1-isopropyl-4-methylenebicyclo[3.1.0]hexan-2-one.
Sigma-Aldrich
(-)-Umbellulone, ≥98% (HPLC)
Pricing and availability is not currently available.