MilliporeSigma
  • Protease-catalyzed monoacylation of 2-O-α-D-glucopyranosyl-L-ascorbic acid in three solvent systems.
Protease-catalyzed monoacylation of 2-O-α-D-glucopyranosyl-L-ascorbic acid in three solvent systems.
Bioscience, biotechnology, and biochemistry (2010-09-14)
Akihiro Tai, Yuji Iwaoka, Tasuku Mori, Hideyuki Ito
ABSTRACT
6-O-dodecanoyl-2-O-α-D-glucopyranosyl-L-ascorbic acid (6-sDode-AA-2G) was synthesized from 2-O-α-D-glucopyranosyl-L-ascorbic acid and vinyl laurate with a protease from Bacillus subtilis in 30% dimethylformamide (DMF)/dioxane with a low water content. The addition of 3% (v/v) water to DMF/dioxane dramatically enhanced the 6-sDode-AA-2G synthesis. The optimum reaction conditions enabled 6-sDode-AA-2G to be synthesized in a yield of 38.1%.
Product Description
Supelco
Ascorbyl Glucoside, Pharmaceutical Secondary Standard; Certified Reference Material