MilliporeSigma
10-year-old girl with severe edema caused by adder bite.
Klinische Padiatrie (2010-09-11)
C Dopfer, F Weigel, G Liebermann, C Vilser, J F Beck, A Dost
ABSTRACT
We report a case of acute hemorrhagic edema in a 10-year-old girl caused by adder bite. Therapy consisted of analgesics, antihistamines, corticosteroids. Antivenum serum was applied in a single dose due to severe spreading of the edema. The patient recovered almost completely after 10 days of treatment.
Product Description
Dimetindene maleate, European Pharmacopoeia (EP) Reference Standard