MilliporeSigma
  • CD203c-based basophil activation test in allergy diagnosis: characteristics and differences to CD63 upregulation.
CD203c-based basophil activation test in allergy diagnosis: characteristics and differences to CD63 upregulation.
Cytometry. Part B, Clinical cytometry (2010-06-10)
Eva M Sturm, Bettina Kranzelbinder, Akos Heinemann, Andrea Groselj-Strele, Werner Aberer, Gunter J Sturm
ABSTRACT
The basophil activation test (BAT) based on CD203c upregulation has been validated as a reliable tool for the diagnosis of IgE-mediated allergies. Nevertheless, CD203c-based BAT is hardly comparable with that of CD63-based tests, as the mechanisms of CD203c versus CD63 induction differ considerably. The aim of the present study was to identify potent influencing factors of the CD203c-based BAT and to emphasize differences between CD63 and CD203c detection. CD203c-based BAT was determined in 82 healthy controls and in 79 allergic patients. The effects of interleukin (IL)-3 and degranulation enhancing substances were investigated and compared with CD63 upregulation. Furthermore, the influence of different storage conditions and incubation times was evaluated and the impact of antiallergic drugs on the test results was assessed. CD203c and CD63 expression was rapidly upregulated reaching a maximum after 20-30 min. Basophil CD203c upregulation assayed after storage times up to 48 h declined already after 4 h. IL-3 treatment increased CD203c and CD63 baseline levels and decreased basophil CD203c responses in a dose-dependent manner. In contrast, cytochalasin B and latrunculin B did not affect CD203c responses but decreased CD63-based BAT. Finally, therapeutic concentrations of dimetindene and desloratadine did not affect CD203c upregulation. CD203c-based basophil activation test should be performed preferentially within 4 h after taking the blood samples. Priming and degranulation-enhancing factors are not required for CD203c-based BAT. In contrast to skin testing, CD203c-based BAT can be performed in patients undergoing antiallergic treatment. © 2010 International Clinical Cytometry Society.
Product Description
Dimetindene maleate, European Pharmacopoeia (EP) Reference Standard