MilliporeSigma
  • Mechanism of carteolol-induced cytosolic Ca2+ mobilization in cultured vascular smooth muscle cells.
Mechanism of carteolol-induced cytosolic Ca2+ mobilization in cultured vascular smooth muscle cells.
Vascular pharmacology (2010-04-27)
Kwou-Yeung Wu, Cheng-Hsien Chang, Hwei-Zu Wang, Show-Jen Hong
ABSTRACT
An increase in cytosolic calcium concentration triggers intracellular signal transduction in vascular cells, which then regulates the vascular contraction. In the present study, the regulatory mechanism of carteolol on the intracellular free Ca(2+) ([Ca(2+)](i)) mobilization was investigated in cultured A7r5 vascular smooth muscle cells. The A7r5 cells were cultured and loaded with fura-2-AM, which was used as a Ca(2+) sensitive fluorescent probe. In both the presence and absence of external Ca(2+), carteolol increased [Ca(2+)](i) with a dose-dependent manner in A7r5 cells at concentrations between 608 microM and 6.08 microM. In a Ca(2+)-containing buffer, carteolol-induced [Ca(2+)](i) showed an initial peak followed by a secondary and persistent plateau. Pretreatment of the cells with La(3+), the plasma membrane Ca(2+) pump inhibitor, and nifedipine, a L-type Ca(2+) channel inhibitor, both partially restrained the carteolol-induced initial peak in [Ca(2+)](i) by 92% and 86%, respectively. Pretreatment of the cells with adrenoceptor antagonists, prazosin inhibited the [Ca(2+)](i) response by 80%, and propranolol enhanced the response by 61%. In the Ca(2+-)-free buffer, pretreatment of the cells with carteolol inhibited the endoplasmic reticulum Ca(2+) pump inhibitor of thapsigargin-induced [Ca(2+)](i) increase by 97%. Pretreatment of the cells with thapsigargin also inhibited the carteolol-induced [Ca(2+)](i) rise by 98%. The internal Ca(2+) release induced by the carteolol was partially inhibited by U73122 (phospholipase C inhibitor) and aristolochic acid, quinacrine (phospholipase A(2) inhibitors). After incubation of carteolol in the Ca(2+)-free buffer, the addition of CaCl(2) increased the Ca(2+) influx, implying that the release of Ca(2+) from internal stores further induced capacitative Ca(2+) entry. These results suggest that carteolol-induced [Ca(2+)](i) increase is mediated by the initial influx via the alpha(1)-adrenoceptor, L-type Ca(2+) channel, nonselective calcium entry channels and release of Ca(2+) from an intracellular store, which is mainly in the endoplasmic reticulum followed by capacitative Ca(2+) entry but decrease via the beta(2)-adrenoceptor. The intracellular Ca(2+) release was also modulated by phospholipase A(2), C-coupled events.
Product Description
Carteolol for system suitability, European Pharmacopoeia (EP) Reference Standard