MilliporeSigma
  • Phenylethanoid and lignan glycosides from polar extracts of Lantana, a genus of verbenaceous plants widely used in traditional herbal therapies.
Phenylethanoid and lignan glycosides from polar extracts of Lantana, a genus of verbenaceous plants widely used in traditional herbal therapies.
Journal of natural products (2009-06-03)
José G Sena Filho, Susan L Nimmo, Haroudo S Xavier, José M Barbosa-Filho, Robert H Cichewicz
ABSTRACT
Many references to the use of Lantana spp. can be found in the ethnopharmacological literature from locations around the globe. This study was focused on examining constituents from the polar extracts of Lantana radula Sw. and Lantana canescens Kunth, for which no prior chemical investigations had been reported. A new phenylethanoid glycoside, raduloside, and lignan glycoside, radulignan, were identified along with the known compounds alyssonoside, arenarioside, calceolarioside E, isonuomioside, samioside, and verbascoside.
Sigma-Aldrich
Calceolarioside A, ≥95% (LC/MS-ELSD)