MilliporeSigma
Polyphenol composition and antioxidant capacity of Epilobium species.
Journal of pharmaceutical and biomedical analysis (2008-11-18)
Barbara Hevesi Tóth, Balázs Blazics, Agnes Kéry
ABSTRACT
Epilobium species (Onagraceae) are commonly used herbal remedies in traditional, adjuvant therapy of benignus prostate hyperplasia (BPH), however the pharmacological and clinical standardization of commercially available Epilobii herba (willow-herb) remains difficult. Willow-herb products usually consist of mixtures from various species, with different phenoloid content, often only partially identified. The present study reports comprehensive LC-MS/MS investigation on the polyphenol composition of the most common Epilobium species, emphasizing the pharmaceutical importance of a uniform standardization protocol in case of their products. The antioxidant capacity of species was evaluated by a simple spectrophotometric method, using ABTS(+) (2,2'azinobis-(3-ethylbenzthiazoline-6-sulfonic acid)). High ratio of macrocyclic tannins, mainly oenothein B was identified in all Epilobium species examined. Flavonoid composition of Epilobium extracts showed several differences, especially comparing E. angustifolium to other species. Myricetin, quercetin, kaempferol and their various glycosides were dominant in samples, but their combination and ratio were distinctive in all cases. Epilobium extracts showed high radical-scavenger activity, comparable to that of well-known antioxidants, Trolox and ascorbic acid. Among species examined, extract of Epilobium parviflorum possessed the highest antioxidant capacity (EC(50)=1.71+/-0.05 microg/ml).
Product Description
Supelco
Oenothein B, analytical standard