MilliporeSigma
  • Direct enantioselective HPLC monitoring of lipase-catalyzed kinetic resolution of tiaprofenic acid in nonstandard HPLC organic solvents.
Direct enantioselective HPLC monitoring of lipase-catalyzed kinetic resolution of tiaprofenic acid in nonstandard HPLC organic solvents.
Chirality (2008-02-05)
Ashraf Ghanem, Mohammed Nabil Aboul-Enein, Aida El-Azzouny, Mohammed F El-Behairy, Ebtessam Al-Humaidi, Alwaleed A Alaidan, Kamilia Amin, Mohammed N Al-Ahdal
ABSTRACT
The first straightforward lipase-catalyzed enantioselective access to enantiomerically enriched tiaprofenic acid as a versatile method in chiral separation of racemates is demonstrated. The latter was directly monitored by enantioselective HPLC using a 3,5-dimethylphenylcarbamate derivative of cellulose-based chiral stationary phase namely Chiralpak IB (the immobilized version of Chiralcel OD). Non-standard HPLC organic solvents were used as diluent to dissolve the "difficult to dissolve" enzyme substrate (the acid) and as eluent for the simultaneous enantioselective HPLC baseline separation of both substrate and product in one run without any further derivatization. The existence of a non-standard HPLC organic solvent (e.g., methyl tert-butyl ether) in the mobile phase composition is mandatory to accomplish the simultaneous enantioselective HPLC baseline separation of both substrate and product.
Product Description
Tiaprofenic acid, European Pharmacopoeia (EP) Reference Standard